Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a15
Entrez Gene ID
365408
Gene Name
membrane-spanning 4-domains, subfamily A, member 15
Gene Aliases
RGD1561158
Chromosome Location
Chr.1: 227033796 - 227048718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191897.1 NP_001178826.1 6 - 7 628 65
XM_006231063.2 XP_006231125.1 6 - 7 2120 65
XM_008760241.1 XP_008758463.1 6 - 7 2134 65

View Full Product Details