Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif3l
Entrez Gene ID
300069
Gene Name
eukaryotic translation initiation factor 3, subunit L
Gene Aliases
Eif3s6ip
Chromosome Location
Chr.7: 120320547 - 120331596 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ209474.1 - - 177 71

View Full Product Details