Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4galnt1
Entrez Gene ID
64828
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 1
Gene Aliases
GalNAc-T, Galgt1
Chromosome Location
Chr.7: 70449728 - 70459559 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022860.2 NP_074051.1 3 - 4 525 148
XM_006241497.2 XP_006241559.1 3 - 4 1052 148
XM_008765419.1 XP_008763641.1 3 - 4 2881 148
XM_008765420.1 XP_008763642.1 2 - 3 1202 148
GenBank mRNA BC081799.1 - 3 - 4 525 148
D17809.1 - 3 - 4 414 148

View Full Product Details