The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130558.1 | NP_001124030.1 | 11 - 12 | 1115 | 66 |
XM_006241486.2 | XP_006241548.1 | 11 - 12 | 1628 | 66 | |
XM_006241488.2 | XP_006241550.1 | 10 - 11 | 1560 | 66 | |
XM_008765415.1 | XP_008763637.1 | 11 - 12 | 1050 | 66 | |
GenBank mRNA | FQ216646.1 | - | 10 - 11 | 939 | 66 |
FQ224725.1 | - | 1 - 2 | 100 | 66 |