Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myf5
Entrez Gene ID
299766
Gene Name
myogenic factor 5
Gene Aliases
-
Chromosome Location
Chr.7: 49729533 - 49732764 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106783.1 NP_001100253.1 2 - 3 809 135

View Full Product Details