Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35e3
Entrez Gene ID
362883
Gene Name
solute carrier family 35, member E3
Gene Aliases
RGD1564876
Chromosome Location
Chr.7: 60765077 - 60778136 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134687.1 NP_001128159.1 3 - 4 841 82
GenBank mRNA BC166468.1 - 3 - 4 841 82
FQ223116.1 - 3 - 4 856 82

View Full Product Details