Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm4
Entrez Gene ID
170820
Gene Name
PR domain containing 4
Gene Aliases
SC-1
Chromosome Location
Chr.7: 23959929 - 23986535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133312.1 NP_579846.1 2 - 3 320 136
XM_006241160.2 XP_006241222.1 4 - 5 782 136
XM_006241161.1 XP_006241223.1 4 - 5 781 136
XM_008765213.1 XP_008763435.1 4 - 5 787 136
GenBank mRNA AF176023.1 - 2 - 3 320 136

View Full Product Details