Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcn
Entrez Gene ID
29139
Gene Name
decorin
Gene Aliases
-
Chromosome Location
Chr.7: 38742250 - 38782282 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024129.1 NP_077043.1 3 - 4 364 134
XM_006241285.1 XP_006241347.1 3 - 4 549 134
GenBank mRNA BC083750.1 - 3 - 4 364 134
FQ210569.1 - 3 - 4 379 134
FQ219808.1 - 3 - 4 397 134
FQ220160.1 - 3 - 4 388 134
FQ220209.1 - 3 - 4 430 134
FQ222189.1 - 3 - 4 433 134
FQ223184.1 - 3 - 4 438 134
FQ228702.1 - 3 - 4 388 134
FQ229041.1 - 3 - 4 452 134
FQ229786.1 - 3 - 4 433 134
X59859.1 - 2 - 3 284 134
Z12298.1 - 3 - 4 493 134

View Full Product Details