Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d30
Entrez Gene ID
299824
Gene Name
TBC1 domain family, member 30
Gene Aliases
RGD1305255
Chromosome Location
Chr.7: 63361443 - 63409250 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241423.2 XP_006241485.1 1 - 2 2167 100

View Full Product Details