The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191987.1 | NP_001178916.1 | 4 - 5 | 540 | 60 |
XM_006241232.2 | XP_006241294.1 | 4 - 5 | 2198 | 60 |