Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab21
Entrez Gene ID
299799
Gene Name
RAB21, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 58261985 - 58286770 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004238.1 NP_001004238.1 2 - 3 436 102
GenBank mRNA BC079323.1 - 2 - 3 436 102

View Full Product Details