Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpihbp1
Entrez Gene ID
300027
Gene Name
glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1
Gene Aliases
RGD1564237
Chromosome Location
Chr.7: 116632496 - 116635543 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130547.1 NP_001124019.1 2 - 3 230 75

View Full Product Details