The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133599.1 | NP_598283.1 | 3 - 4 | 284 | 86 |
GenBank mRNA | AB001075.1 | - | 3 - 4 | 284 | 86 |