The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024371.1 | NP_001019542.1 | 3 - 4 | 650 | 117 |
XM_008765456.1 | XP_008763678.1 | 4 - 5 | 640 | 117 | |
GenBank mRNA | BC087105.1 | - | 3 - 4 | 650 | 117 |
FQ214436.1 | - | - | 659 | 117 |