Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ebag9
Entrez Gene ID
299864
Gene Name
estrogen receptor binding site associated, antigen, 9
Gene Aliases
-
Chromosome Location
Chr.7: 83564563 - 83582516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009665.1 NP_001009665.1 2 - 3 294 137
XM_006241613.2 XP_006241675.1 2 - 3 669 137
XM_008765451.1 XP_008763673.1 1 - 2 382 137
XM_008765452.1 XP_008763674.1 2 - 3 3332 137
GenBank mRNA BC087093.1 - 2 - 3 294 137

View Full Product Details