Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo32
Entrez Gene ID
171043
Gene Name
F-box protein 32
Gene Aliases
Atrogin1, MAFbx
Chromosome Location
Chr.7: 98065664 - 98098071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133521.1 NP_598205.1 7 - 8 818 71
GenBank mRNA AY059628.1 - 7 - 8 818 71

View Full Product Details