The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006241900.2 | XP_006241962.1 | 4 - 5 | 541 | 74 |
XM_006241901.2 | XP_006241963.1 | 4 - 5 | 542 | 74 | |
XM_006241902.1 | XP_006241964.1 | 2 - 3 | 155 | 74 | |
XM_008765628.1 | XP_008763850.1 | 4 - 5 | 1930 | 74 | |
XM_216954.8 | XP_216954.4 | 4 - 5 | 540 | 74 |