Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtpbp1
Entrez Gene ID
300077
Gene Name
GTP binding protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 120923274 - 120947723 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199315.1 NP_001186244.1 9 - 10 1578 61
GenBank mRNA GU256773.1 - 9 - 10 1538 61

View Full Product Details