The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126269.1 | NP_001119741.1 | 4 - 5 | 449 | 63 |
GenBank mRNA | BC166457.1 | - | 4 - 5 | 344 | 63 |