The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126269.1 | NP_001119741.1 | 8 - 9 | 896 | 78 |
GenBank mRNA | BC166457.1 | - | 8 - 9 | 791 | 78 |