Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h3
Entrez Gene ID
300032
Gene Name
zinc finger CCCH type containing 3
Gene Aliases
Zc3hdc3
Chromosome Location
Chr.7: 116787762 - 116872558 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134865.1 NP_001128337.1 11 - 12 2863 110

View Full Product Details