Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tssk5
Entrez Gene ID
315095
Gene Name
testis-specific serine kinase 5
Gene Aliases
RGD1308613
Chromosome Location
Chr.7: 117437304 - 117440342 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014086.2 NP_001014108.2 5 - 6 950 94
GenBank mRNA BC089206.1 - 4 - 5 1119 94

View Full Product Details