Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf21b
Entrez Gene ID
300117
Gene Name
PHD finger protein 21B
Gene Aliases
RGD1308739
Chromosome Location
Chr.7: 125649688 - 125720694 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130680.1 NP_001124152.1 12 - 13 1653 92
XM_008765685.1 XP_008763907.1 12 - 13 1240 92

View Full Product Details