The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134495.1 | NP_001127967.1 | 10 - 11 | 1164 | 103 |
XM_006241631.2 | XP_006241693.1 | 14 - 15 | 1828 | 103 |