The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012991.1 | NP_037123.1 | 7 - 8 | 1524 | 115 |
GenBank mRNA | BC097337.1 | - | 7 - 8 | 1242 | 115 |
U41845.2 | - | 7 - 8 | 1524 | 115 |