Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc14
Entrez Gene ID
500900
Gene Name
leucine rich repeat containing 14
Gene Aliases
-
Chromosome Location
Chr.7: 117773218 - 117779520 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024354.1 NP_001019525.1 2 - 3 502 66
XM_006241860.2 XP_006241922.1 2 - 3 757 66
XM_006241861.1 XP_006241923.1 2 - 3 390 66
XM_006241862.2 XP_006241924.1 3 - 4 1275 66
XM_006241865.1 XP_006241927.1 3 - 4 485 66
GenBank mRNA BC092583.1 - 2 - 3 502 66

View Full Product Details