Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nab2
Entrez Gene ID
314910
Gene Name
Ngfi-A binding protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 70963580 - 70971480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241459.2 XP_006241521.1 5 - 6 2985 115

View Full Product Details