Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stim1
Entrez Gene ID
361618
Gene Name
stromal interaction molecule 1
Gene Aliases
-
Chromosome Location
Chr.1: 167373894 - 167533412 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108496.2 NP_001101966.2 4 - 5 873 68
GenBank mRNA BC166914.1 - 4 - 5 532 68

View Full Product Details