Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyrm1
Entrez Gene ID
365361
Gene Name
LYR motif containing 1
Gene Aliases
RGD1563498
Chromosome Location
Chr.1: 189663200 - 189685842 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276487.1 NP_001263416.1 1 - 2 63 77
GenBank mRNA BC166909.1 - 1 - 2 60 77

View Full Product Details