The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009646.1 | NP_001009646.1 | 2 - 3 | 601 | 108 |
XM_006230246.2 | XP_006230308.1 | 2 - 3 | 772 | 108 | |
XM_008759877.1 | XP_008758099.1 | 2 - 3 | 1077 | 108 | |
XM_008759878.1 | XP_008758100.1 | 3 - 4 | 864 | 108 | |
GenBank mRNA | BC088177.1 | - | 2 - 3 | 601 | 108 |
FQ209866.1 | - | 2 - 3 | 595 | 108 |