Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Omp
- Entrez Gene ID
-
24612
- Gene Name
- olfactory marker protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 163097159 - 163097709 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M15644.1 |
- |
- |
2083 |
75 |
View Full Product Details