Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Umod
Entrez Gene ID
25128
Gene Name
uromodulin
Gene Aliases
THP
Chromosome Location
Chr.1: 189186027 - 189199939 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017082.1 NP_058778.1 2 - 3 896 131
XM_006230107.2 XP_006230169.1 3 - 4 1018 131
GenBank mRNA BC081814.1 - 2 - 3 902 131
M63510.1 - 2 - 3 896 131
S75960.1 - 2 - 3 887 131

View Full Product Details