The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106297.2 | NP_001099767.1 | 5 - 6 | 946 | 122 |
XM_006230251.2 | XP_006230313.1 | 6 - 7 | 2708 | 122 | |
XM_006230252.2 | XP_006230314.1 | 6 - 7 | 2716 | 122 | |
XM_008759879.1 | XP_008758101.1 | 6 - 7 | 2722 | 122 | |
GenBank mRNA | BC168174.1 | - | 5 - 6 | 946 | 122 |