The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163164.1 | NP_001156636.1 | 4 - 5 | 752 | 119 |
XM_008759659.1 | XP_008757881.1 | 4 - 5 | 641 | 119 | |
GenBank mRNA | FQ232050.1 | - | 2 - 3 | 211 | 119 |
HQ898855.1 | - | 4 - 5 | 749 | 119 |