Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr1h4
Entrez Gene ID
60351
Gene Name
nuclear receptor subfamily 1, group H, member 4
Gene Aliases
Fxr
Chromosome Location
Chr.7: 30003429 - 30162095 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021745.1 NP_068513.1 9 - 10 1341 67
XM_008765260.1 XP_008763482.1 8 - 9 1711 67
XM_008765261.1 XP_008763483.1 8 - 9 1698 67
XM_008765262.1 XP_008763484.1 9 - 10 1392 67
GenBank mRNA BC083815.1 - 8 - 9 1219 67
U18374.1 - 9 - 10 1341 67

View Full Product Details