The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108743.1 | NP_001102213.1 | 1 - 2 | 204 | 88 |
XM_006241265.2 | XP_006241327.1 | 2 - 3 | 609 | 88 |