Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Best3
Entrez Gene ID
314847
Gene Name
bestrophin 3
Gene Aliases
Vmd2l3
Chromosome Location
Chr.7: 59971145 - 60057162 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191783.1 NP_001178712.1 5 - 6 715 112
XM_008765394.1 XP_008763616.1 7 - 8 1859 112
XM_008765395.1 XP_008763617.1 5 - 6 1596 112
GenBank mRNA FQ222375.1 - 7 - 8 1210 112

View Full Product Details