Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amdhd1
Entrez Gene ID
299735
Gene Name
amidohydrolase domain containing 1
Gene Aliases
RGD1308879
Chromosome Location
Chr.7: 34362635 - 34406282 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191781.1 NP_001178710.1 3 - 4 320 62

View Full Product Details