Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d10b
Entrez Gene ID
365372
Gene Name
TBC1 domain family, member 10b
Gene Aliases
RGD1309191
Chromosome Location
Chr.1: 198641018 - 198652674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108921.1 NP_001102391.1 6 - 7 1459 97

View Full Product Details