Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Me3
Entrez Gene ID
361602
Gene Name
malic enzyme 3, NADP(+)-dependent, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.1: 153861569 - 154056594 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108491.2 NP_001101961.2 2 - 3 260 119
XM_006229666.1 XP_006229728.1 2 - 3 260 119

View Full Product Details