The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009239.1 | NP_001009239.1 | 3 - 4 | 265 | 74 |
GenBank mRNA | BC088450.1 | - | 3 - 4 | 265 | 74 |
X51536.1 | - | 3 - 4 | 271 | 74 |