The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191838.1 | NP_001178767.1 | 13 - 14 | 1289 | 61 |
GenBank mRNA | BC099160.1 | - | 4 - 5 | 277 | 61 |