Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppme1
Entrez Gene ID
361613
Gene Name
protein phosphatase methylesterase 1
Gene Aliases
PME-1, RGD1309683
Chromosome Location
Chr.1: 165335242 - 165382216 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191838.1 NP_001178767.1 3 - 4 430 90

View Full Product Details