Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bai3
Entrez Gene ID
301309
Gene Name
brain-specific angiogenesis inhibitor 3
Gene Aliases
-
Chromosome Location
Chr.9: 31280623 - 32022098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106898.1 NP_001100368.1 21 - 22 3245 63
XM_008766928.1 XP_008765150.1 5 - 6 936 63
XM_008766929.1 XP_008765151.1 6 - 7 829 63
XM_008766930.1 XP_008765152.1 6 - 7 2250 63
XM_008766931.1 XP_008765153.1 6 - 7 1066 63
XM_008766932.1 XP_008765154.1 5 - 6 936 63

View Full Product Details