Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bai3
Entrez Gene ID
301309
Gene Name
brain-specific angiogenesis inhibitor 3
Gene Aliases
-
Chromosome Location
Chr.9: 31280623 - 32022098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106898.1 NP_001100368.1 7 - 8 1741 95

View Full Product Details