Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crisp1
Entrez Gene ID
64827
Gene Name
cysteine-rich secretory protein 1
Gene Aliases
Aeg, Crisp-1
Chromosome Location
Chr.9: 24047454 - 24088064 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022859.1 NP_074050.1 4 - 5 308 73
GenBank mRNA M31173.1 - 4 - 5 308 73
X04643.1 - 4 - 5 280 73

View Full Product Details