Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dlk2
Entrez Gene ID
316232
Gene Name
delta-like 2 homolog (Drosophila)
Gene Aliases
DLK-2, Egfl9
Chromosome Location
Chr.9: 17061187 - 17065600 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108202.1 NP_001101672.1 5 - 6 540 135
XM_006244523.2 XP_006244585.1 5 - 6 507 135
XM_008766865.1 XP_008765087.1 5 - 6 774 135

View Full Product Details