The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198756.2 | NP_942051.1 | 5 - 6 | 443 | 80 |
GenBank mRNA | BC058507.1 | - | 5 - 6 | 447 | 80 |
FQ215521.1 | - | 6 - 7 | 622 | 80 |