The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108203.1 | NP_001101673.1 | 11 - 12 | 1050 | 80 |
XM_006244524.2 | XP_006244586.1 | 11 - 12 | 1046 | 80 | |
XM_006244525.2 | XP_006244587.1 | 11 - 12 | 1043 | 80 | |
XM_006244526.2 | XP_006244588.1 | 10 - 11 | 916 | 80 | |
XM_006244527.2 | XP_006244589.1 | 10 - 11 | 948 | 80 | |
XM_006244528.2 | XP_006244590.1 | 10 - 11 | 945 | 80 | |
XM_006244529.2 | XP_006244591.1 | 9 - 10 | 818 | 80 | |
XM_006244530.2 | XP_006244592.1 | 9 - 10 | 815 | 80 | |
XM_006244532.2 | XP_006244594.1 | 10 - 11 | 943 | 80 | |
XM_006244533.2 | XP_006244595.1 | 9 - 10 | 813 | 80 | |
XM_006244534.2 | XP_006244596.1 | 10 - 11 | 1016 | 80 | |
XM_006244535.2 | XP_006244597.1 | 8 - 9 | 1311 | 80 |