Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cops2
Entrez Gene ID
261736
Gene Name
COP9 signalosome subunit 2
Gene Aliases
CSN2, Trip15
Chromosome Location
Chr.3: 118109394 - 118137125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153297.1 NP_695209.1 5 - 6 614 135
XM_006234901.2 XP_006234963.1 5 - 6 564 135
XM_008762146.1 XP_008760368.1 5 - 6 564 135
GenBank mRNA AB081072.1 - 5 - 6 614 135
BC061864.1 - 5 - 6 535 135

View Full Product Details